missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade, Thermo Scientific Chemicals

Catalog Number 10565291
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
80526-82-5
C4H13NO4S
171.21
MFCD00036149
DWTYPCUOWWOADE-UHFFFAOYSA-M
tetramethylammonium hydrogen sulfate, tetramethylammonium bisulfate, tetramethylammonium hydrogen sulphate, tetramethylammonium hydrogensulfate, hydrogen sulfate; tetramethylazanium, methanaminium, n,n,n-trimethyl-, sulfate 1:1, hydrogen sulfate; tetramethylammonium, acmc-209pjn, ch3 4n hso4 .h2o, tetramethylammonium bisulfate solution
157340
hydrogen sulfate;tetramethylazanium
C[N+](C)(C)C.OS(=O)(=O)[O-]
80526-82-5
C4H13NO4S
171.21
MFCD00036149
DWTYPCUOWWOADE-UHFFFAOYSA-M
tetramethylammonium hydrogen sulfate, tetramethylammonium bisulfate, tetramethylammonium hydrogen sulphate, tetramethylammonium hydrogensulfate, hydrogen sulfate; tetramethylazanium, methanaminium, n,n,n-trimethyl-, sulfate 1:1, hydrogen sulfate; tetramethylammonium, acmc-209pjn, ch3 4n hso4 .h2o, tetramethylammonium bisulfate solution
157340
hydrogen sulfate;tetramethylazanium
C[N+](C)(C)C.OS(=O)(=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Absorbance (0.005M in water), (0.005 M in water) at 0nm, 0.05 max. at 220nm, 0.15 max. at 200nm
Melting Point 295.0°C to 297.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 171.21
Percent Purity 99+%
Grade HPLC
Physical Form Powder
Chemical Name or Material Tetramethylammonium hydrogensulfate
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hydrogensulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'