missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific™

Catalog Number p-3758798
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
558-32-7
C4H12F6NP
219.111
MFCD00012012
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
11209
tetramethylazanium;hexafluorophosphate
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
This item is not returnable. View return policy
558-32-7
C4H12F6NP
219.111
MFCD00012012
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
11209
tetramethylazanium;hexafluorophosphate
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)4NPF6
Refractive Index 1.4820 to 1.4850
Quantity 25 g
Formula Weight 219.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'