missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium formate, 30% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Tetramethylammonium formate may provide the lowest temperature sensitivity, additionally reducing corrosion of stainless-steel flow system by chloride-induced depassivation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linear Formula | (CH3)4N(COOH) |
Quantity | 100 mL |
Beilstein | 3916177 |
Solubility Information | Fully miscible in water. |
Formula Weight | 119.16 |
Concentration or Composition (by Analyte or Components) | 30% w/w aq. soln. |
Physical Form | Liquid |
Chemical Name or Material | Tetramethylammonium formate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'