missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals

Catalog Number p-4509862
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
681-84-5
C4H12O4Si
152.22
MFCD00008341
LFQCEHFDDXELDD-UHFFFAOYSA-N
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
12682
tetramethyl silicate
CO[Si](OC)(OC)OC
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
681-84-5
C4H12O4Si
152.22
MFCD00008341
LFQCEHFDDXELDD-UHFFFAOYSA-N
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
12682
tetramethyl silicate
CO[Si](OC)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2.0°C
Density 1.0300g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Si(OCH3)4
Refractive Index 1.3670 to 1.3700
Quantity 25 g
Beilstein 01,287
Specific Gravity 1.03
Solubility Information Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol
Viscosity 0.7 mPa.s (20°C)
Formula Weight 152.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetramethyl orthosilicate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethyl orthosilicate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'