missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(triphenylphosphine)nickel(0), Thermo Scientific Chemicals

Catalog Number p-7049962
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
15133-82-1
C72H60NiP4
1107.86
MFCD00010011
KFBKRCXOTTUAFS-UHFFFAOYSA-N
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
11979963
nickel;triphenylphosphane
[Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
15133-82-1
C72H60NiP4
1107.86
MFCD00010011
KFBKRCXOTTUAFS-UHFFFAOYSA-N
ni pph3 4, tetrakis triphenylphosphine nickel, tetrakis triphenylphosphine nickel 0, nickel-tetrakis triphenylphosphine, tetrakis triphenylphosphine nickel 0 , ni 4-7 % approx.
11979963
nickel;triphenylphosphane
[Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Tetrakis(triphenylphosphine)nickel(0) is a catalyst for coupling reactions of organic halides, organometallic compounds and also for the cyclooligomerization of cumulenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 123°C to 128°C
Color Brown
Linear Formula Ni[P(C6H5)3]4
Quantity 250 mg
UN Number UN1325
Sensitivity Air & Moisture Sensitive
Formula Weight 1107.83
Physical Form Crystalline
Chemical Name or Material Tetrakis(triphenylphosphine)nickel(0)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'