missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058644
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3275-24-9
C8H24N4Ti
224.18
MFCD00014861
MNWRORMXBIWXCI-UHFFFAOYSA-N
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
123185
dimethylazanide;titanium(4+)
[Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3275-24-9
C8H24N4Ti
224.18
MFCD00014861
MNWRORMXBIWXCI-UHFFFAOYSA-N
tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
123185
dimethylazanide;titanium(4+)
[Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown-Yellow
Density 0.96 g/mL
Boiling Point 50°C (0.5 mmHg)
Flash Point −30°C (−22°F)
Linear Formula Ti[N(CH3)2]4
Quantity 5 g
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 224.19
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'