missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051819
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
18865-74-2
C44H80O8Zr
828.34
MFCD00145380,MFCD00145380
ANBXKEDJQLYCLX-UHFFFAOYSA-N
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
50919870
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
This item is not returnable. View return policy
18865-74-2
C44H80O8Zr
828.34
MFCD00145380,MFCD00145380
ANBXKEDJQLYCLX-UHFFFAOYSA-N
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
50919870
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C

MOCVD of zirconium

MOCVD of zirconium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 318°C to 320°C
Boiling Point 180°C (0.1 mmHg)(sublimation)
Quantity 1 g
Formula Weight 824.3
Percent Purity 99.99%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'