missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraisobutyldialuminoxane, 10 wt.% solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Color | Colorless |
Density | 0.8670g/mL |
Boiling Point | 111°C |
Flash Point | -8°C |
Packaging | AcroSeal™ Glass Bottle |
Quantity | 100 mL |
Specific Gravity | 0.867 |
Formula Weight | 298.42 |
Physical Form | Solution |
Chemical Name or Material | Tetraisobutyldialuminoxane |
Safety and Handling
Product Identifier
- Tetraisobutyldialuminoxane, 10 wt.% solution in toluene
- Danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'