Learn More
Thermo Scientific Maybridge Tetrahydrothiopyran-4-carbonitrile, 10g, 97%
Brand: Thermo Scientific Maybridge CC61916EA
Tetrahydrothiopyran-4-carbonitrile, 10g, 97%
Code : 31
Additional Details : CAS Number : 195503-40-3 Weight : 0.06000kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III