missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophen-3-one, 97%, Thermo Scientific Chemicals

Catalog Number p-7081154
Click to view available options
Quantity:
5 g
10 g
25 g
250 g
This item is not returnable. View return policy
1003-04-9
C4H6OS
102.15
MFCD00005412
DSXFPRKPFJRPIB-UHFFFAOYSA-N
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
61252
thiolan-3-one
O=C1CCSC1
This item is not returnable. View return policy
1003-04-9
C4H6OS
102.15
MFCD00005412
DSXFPRKPFJRPIB-UHFFFAOYSA-N
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
61252
thiolan-3-one
O=C1CCSC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.192
Boiling Point 174°C to 175°C
Flash Point 77°C (170°F)
Odor Characteristic
Refractive Index 1.529
Quantity 5 g
Beilstein 105201
Formula Weight 102.16
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophen-3-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'