missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-4-ylacetic acid, 97%, Thermo Scientific™

Catalog Number p-3830492
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
85064-61-5
C7H11O3
143.16
MFCD01631204
PBXYNWPYMVWJAH-UHFFFAOYSA-M
tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid
2773575
2-(oxan-4-yl)acetic acid
[O-]C(=O)CC1CCOCC1
This item is not returnable. View return policy
85064-61-5
C7H11O3
143.16
MFCD01631204
PBXYNWPYMVWJAH-UHFFFAOYSA-M
tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid
2773575
2-(oxan-4-yl)acetic acid
[O-]C(=O)CC1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-4-ylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'