Learn More
Thermo Scientific Maybridge Tetrahydropyran-2-carbonyl chloride, 1g, 95%
Brand: Thermo Scientific Maybridge CC41202DA
Tetrahydropyran-2-carbonyl chloride, 1g, 95%
Code : 31
Additional Details : CAS Number : 40053-81-4 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 8 Pack group : II