missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfurylamine 97%, ACROS Organics™

Brand:  Acros Organics 157821000

Code : 29

Additional Details : CAS Number : 4795-29-3 Weight : 0.15000kg Transport : UN number : 2943 Chem class : 1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10754471

  • £13.80 / 100g


Estimated Shipment
14-02-2018
Add to basket


Specifications

Chemical Identifiers

Tetrahydrofurfurylamine
4795-29-3
97%
101.15
2-(Aminomethyl)tetrahydrofuran
97%
97%
C5H11NO
MFCD00005373

Specifications

Tetrahydrofurfurylamine
45°C
153.0° to 154.0°C (744.0 mmHg)
100g
4795-29-3
Glass bottle
Solubility in water: miscible with water. Other solubilities: soluble in alcohol and ether
97%
C5H11NO
MFCD00005373
2-(Aminomethyl)tetrahydrofuran
0.9800g/mL
0.98
Clear colorless to light yellow
97%
Authentic
1.4540 to 1.4560
Liquid
97%
101.15
18, II, 415
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrahydrofurfurylamine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS