missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number p-3830435
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
97-99-4
C5H10O2
102.13
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYNA-N
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
7360
OCC1CCCO1
This item is not returnable. View return policy
97-99-4
C5H10O2
102.13
MFCD00005372
BSYVTEYKTMYBMK-UHFFFAOYNA-N
tetrahydrofurfuryl alcohol, tetrahydro-2-furanmethanol, tetrahydrofuran-2-yl methanol, thfa, 2-furanmethanol, tetrahydro, tetrahydro-2-furanylmethanol, qo thfa, tetrahydro-2-furancarbinol, tetrahydrofuryl carbinol, furfuryl alcohol, tetrahydro
7360
OCC1CCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -80.0°C
Density 1.0543g/mL
Boiling Point 178.0°C
Quantity 1 L
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4510 to 1.4530
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.0543
Viscosity 6.24 mPa.s (20°C)
Beilstein 17, V,3, 115
Merck Index 15, 9358
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, ether, acetone, chloroform, and benzene
IUPAC Name (oxolan-2-yl)methanol
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl alcohol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'