missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, Extra Pure, SLR, Stabilised with 0.025% BHT, Fisher Chemical™
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
39.80 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
39.80 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal can
Plastic-coated Glass Bottle
Description
C4H8O, CAS Number-109-99-9, oxacyclopentane, tetraidrofurano, 1,4-epoxybutane, furan, tetrahydro, butylene oxide, hydrofuran, tetrahydrofuranne, tetrahydrofuran, furanidine, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C
Specifications
pH | 7 to 8 |
Melting Point | -108.4°C |
Color | Colorless |
Boiling Point | 66°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 200mbar at 20°C |
Viscosity | 0.55cp at 20°C |
Formula Weight | 72.11g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'