missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, Extra Dry, for Synthesis, Unstabilised, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Description
Tetrahydrofuran, Extra Dry, for Synthesis, Unstabilised
Specifications
pH | 7 to 8 |
Melting Point | -108.4°C |
Color | Colorless |
Boiling Point | 66°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 200mbar at 20°C |
Viscosity | 0.55cp at 20°C |
Formula Weight | 72.11g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'