missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-d{8}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog Number 15471457
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Deuterated THF is used as a solvent in many organic reactions and in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.985 g/mL
Flash Point -17°C (-1°F)
Quantity 1 g
CAS 1693-74-9
Melting Point -106°C
Boiling Point 66°C
Molecular Formula C4H8O
MDL Number MFCD00044238
UN Number UN2056
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
Solubility Information Miscible with water.
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
Molecular Weight (g/mol) 80.16
PubChem CID 80290
Formula Weight 80.16
Percent Purity 99.5%
Chemical Name or Material Tetrahydrofuran-d8, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'