missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-d{8}, 100%(Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051978
Click to view available options
Quantity:
10 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1693-74-9
C4H8O
80.16
MFCD00044238
WYURNTSHIVDZCO-SVYQBANQSA-N
tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
80290
2,2,3,3,4,4,5,5-octadeuteriooxolane
[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1693-74-9
C4H8O
80.16
MFCD00044238
WYURNTSHIVDZCO-SVYQBANQSA-N
tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
80290
2,2,3,3,4,4,5,5-octadeuteriooxolane
[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Tetrahydrofuran-d_8 is a deuterated THF that is used in various synthesis reactions and also used as a precursor for various polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.985 g/mL
Flash Point −17°C (−1°F)
Packaging Prescored Ampule
Quantity 10 Ea.
Melting Point -106°C
Boiling Point 66°C
UN Number UN2056
Solubility Information Soluble in water.
Formula Weight 80.16
Percent Purity 100%
Chemical Name or Material Tetrahydrofuran-d8, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'