missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-3-one, 95%, Thermo Scientific Chemicals

Catalog Number p-7057351
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22929-52-8
C4H6O2
86.09
MFCD07778393
JLPJFSCQKHRSQR-UHFFFAOYSA-N
3-oxotetrahydrofuran, dihydrofuran-3 2h-one, dihydro-3 2h-furanone, dihydrofuran-3-one, dihydro-furan-3-one, tetrahydrofuran-3-one, 3 2h-furanone, dihydro, 3-oxo-1,4-epoxybutane, tetrahydrofuran-3-yloxy
529392
oxolan-3-one
C1COCC1=O
This item is not returnable. View return policy
22929-52-8
C4H6O2
86.09
MFCD07778393
JLPJFSCQKHRSQR-UHFFFAOYSA-N
3-oxotetrahydrofuran, dihydrofuran-3 2h-one, dihydro-3 2h-furanone, dihydrofuran-3-one, dihydro-furan-3-one, tetrahydrofuran-3-one, 3 2h-furanone, dihydro, 3-oxo-1,4-epoxybutane, tetrahydrofuran-3-yloxy
529392
oxolan-3-one
C1COCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Physical Form Liquid
Beilstein 105200
Formula Weight 86.09
Percent Purity 95%
Chemical Name or Material Tetrahydrofuran-3-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'