missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-β-carboline, 98%, ACROS Organics™

Brand:  Acros Organics 427580050

Code : 29

Additional Details : CAS Number : 16502-01-5 Weight : 0.05500kg Transport : UN number : 3263 Chem class : 8 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10584954

  • £99.70 / 5g


Call for latest availability
+44 (0)1509 555 500
Add to basket


Specifications

Chemical Identifiers

Tetrahydro-β-carboline
≥97.5% (HPLC)
C11H12N2
MFCD00004954
16502-01-5
98%
172.23
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole

Specifications

Tetrahydro-β-carboline
5g
Authentic
Glass bottle
≥97.5% (HPLC)
C11H12N2
MFCD00004954
White to slightly yellow
16502-01-5
Consistent with structure (1H-NMR)
Powder or Crystals
98%
172.23
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrahydro-beta-carboline

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS