missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-β-carboline, 98%, Thermo Scientific Chemicals

Catalog Number p-4089832
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16502-01-5
C11H13ClN2
208.69
MFCD00004954
PHLJRXUBLWEPCM-UHFFFAOYSA-N
tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
107838
2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
[H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21
This item is not returnable. View return policy
16502-01-5
C11H13ClN2
208.69
MFCD00004954
PHLJRXUBLWEPCM-UHFFFAOYSA-N
tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
107838
2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
[H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 206°C to 208°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Tetrahydro-β-carboline
compliance-icons
Product Identifier
  • Tetrahydro-beta-carboline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'