missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-pyran-4-ol, Thermo Scientific™

Catalog Number 11916201
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2081-44-9
C5H10O2
102.13
LMYJGUNNJIDROI-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-ol, tetrahydro-4-pyranol, 4-hydroxytetrahydropyran, tetrahydro-4h-pyran-4-ol, tetrahydro-pyran-4-ol, 2h-pyran-4-ol, tetrahydro, 4-hydroxytetrahydropyan, tetrahydropyran-4-ol, 4-hydroxytetrahydro-4h-pyran, 4-hydroxytetrahydro-2h-pyran
74956
oxan-4-ol
C1COCCC1O
This item is not returnable. View return policy
2081-44-9
C5H10O2
102.13
LMYJGUNNJIDROI-UHFFFAOYSA-N
tetrahydro-2h-pyran-4-ol, tetrahydro-4-pyranol, 4-hydroxytetrahydropyran, tetrahydro-4h-pyran-4-ol, tetrahydro-pyran-4-ol, 2h-pyran-4-ol, tetrahydro, 4-hydroxytetrahydropyan, tetrahydropyran-4-ol, 4-hydroxytetrahydro-4h-pyran, 4-hydroxytetrahydro-2h-pyran
74956
oxan-4-ol
C1COCCC1O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.0700g/mL
Boiling Point 87.0°C (15.0 mmHg)
Flash Point 190°C
Refractive Index 1.46
Quantity 1 g
Beilstein 17,II,106
Specific Gravity 1.07
Formula Weight 102.13
Chemical Name or Material Tetrahydro-4H-pyran-4-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'