missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrazolidinone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4510585
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1752-88-1
C3H7ClN2O
122.55
MFCD00043787
HAYWTXQRYXYRAF-UHFFFAOYSA-N
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
2733712
[H+].[Cl-].O=C1CCNN1
This item is not returnable. View return policy
1752-88-1
C3H7ClN2O
122.55
MFCD00043787
HAYWTXQRYXYRAF-UHFFFAOYSA-N
3-pyrazolidinone hydrochloride, pyrazolidin-3-one hydrochloride, 4,5-dihydro-1h-pyrazol-3-ol hydrochloride, 3-pyrazolidinone, monohydrochloride, tetrahydro-3h-pyrazol-3-one hydrochloride, 3-pyrazolidinone hcl, 3-pyrazolidone hydrochloride, ksc493k0l, pyrazolidin-3-on-hydrochloride, pyrazolidin-3-one hydrogen chloride
2733712
[H+].[Cl-].O=C1CCNN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
IUPAC Name pyrazolidin-3-one;hydrochloride
Formula Weight 122.55
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Pyrazolidinone hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'