missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-3-furoic acid, 97%, ACROS Organics™

Brand:  Acros Organics 433450250

Code : 29

Additional Details : CAS Number : 89364-31-8 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11946181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Tetrahydro-3-furoic acid
97%
116.12
≥96.0%
C5H8O3

Specifications

Tetrahydro-3-furoic acid
Colorless to pale yellow
89364-31-8
(250ppm BHT inhibitor)
≥96.0%
C5H8O3
1.214g/mL
25g
Authentic
Liquid
97%
116.12
Videos
SDS