missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-3-furoic acid, 97%, ACROS Organics™

Brand:  Acros Organics 433450050

Code : 29

Additional Details : CAS Number : 89364-31-8 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11936181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Tetrahydro-3-furoic acid
97%
116.12
97%
C5H8O3

Specifications

Tetrahydro-3-furoic acid
113°C
138.0° to 140.0°C (12.0 mmHg)
5g
Authentic
Liquid
97%
116.12
1.2140g/mL
1.214
Colorless to pale yellow
89364-31-8
(250ppm BHT inhibitor)
97%
C5H8O3
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrahydro-3-furoic acid, contains 250 ppm BHT as inhibitor

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS