Tetrahydro-2H-thiopyran-4-carbaldehyde, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC61904CB

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 50675-19-9 Weight: 0.05025kg Transport: UN number : 2810 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 250mg

Product Code. 10285424

Quantity Price
1 £ 74.0 / 250mg
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Tetrahydro-2H-thiopyran-4-carbaldehyde
CAS 50675-19-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H10OS
Formula Weight 130.21
MDL Number MFCD03425266

Specification

Formula Weight 130.21
Quantity 250mg
Assay 97%
Chemical Name or Material Tetrahydro-2H-thiopyran-4-carbaldehyde
Molecular Formula C6H10OS
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 100.0
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Assay Percent Range 97%
CAS 50675-19-9
MDL Number MFCD03425266
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.