missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 130.14 |
Percent Purity | 97% |
Chemical Name or Material | Tetrahydro-2H-pyran-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'