missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium Bromide, 99%, ACROS Organics™

£45.91 - £148.00

Chemical Identifiers

CAS 4328-13-6
Molecular Formula C24H52BrN
Molecular Weight (g/mol) 434.58
MDL Number MFCD00011858
InChI Key SYZCZDCAEVUSPM-UHFFFAOYSA-M
Synonym tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
PubChem CID 78026
IUPAC Name tetrahexylazanium;bromide
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[Br-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10055880
SDS View Documents
Acros Organics
420400250
25g Glass bottle
£45.91
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 6 Please to purchase this item. Need a web account? Register with us today!
10464045
SDS View Documents
Acros Organics
420401000
100g Glass bottle
£148.00
100g
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

4328-13-6
434.58
SYZCZDCAEVUSPM-UHFFFAOYSA-M
78026
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[Br-]
C24H52BrN
MFCD00011858
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
tetrahexylazanium;bromide

Specifications

Tetrahexylammonium bromide
Glass bottle
98.0°C to 100.0°C
4328-13-6
C24H52BrN
MFCD00011858
SYZCZDCAEVUSPM-UHFFFAOYSA-M
tetrahexylazanium;bromide
78026
Powder or Flakes
Authentic
White to Yellow
25g
98.5% min. (Argentometry)
[CH3(CH2)5]4NBr
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[Br-]
434.58
434.58
99%
SDS