missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium Bromide, 99%, ACROS Organics™

Brand:  Acros Organics 420400250

Code : 29

Additional Details : CAS Number : 4328-13-6 Weight : 0.07500kg

Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10055880

  • £45.91 / 25g

In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

4328-13-6
434.58
SYZCZDCAEVUSPM-UHFFFAOYSA-M
78026
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[Br-]
C24H52BrN
MFCD00011858
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
tetrahexylazanium;bromide

Specifications

Tetrahexylammonium bromide
Glass bottle
98.0°C to 100.0°C
4328-13-6
C24H52BrN
MFCD00011858
SYZCZDCAEVUSPM-UHFFFAOYSA-M
tetrahexylazanium;bromide
78026
Powder or Flakes
Authentic
White to Yellow
25g
98.5% min. (Argentometry)
[CH3(CH2)5]4NBr
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[Br-]
434.58
434.58
99%
Videos
SDS Documents