Tetraformylhydrazine, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 380480010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 52023-52-6 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10556363

Quantity Price
1 £ 25.1 / 1g
EU Stock 2
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Tetraformylhydrazine
Assay Percent Range 98%
Molecular Formula C4H4N2O4
Linear Formula [N(COH)2]2
Formula Weight 144.09
MDL Number MFCD08059355

Specification

Physical Form Powder
Formula Weight 144.09
Quantity 1g
Color White
Linear Formula [N(COH)2]2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 52023-52-6
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD08059355
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Elemental Analysis 97.5% min.
Packaging Glass bottle
Molecular Formula C4H4N2O4
Chemical Name or Material Tetraformylhydrazine
Documentation

Technical documents

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.