missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyltin, 98%, Thermo Scientific Chemicals

Catalog Number 11304839
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
This item is not returnable. View return policy
597-64-8
C8H20Sn
234.96
MFCD00009020
RWWNQEOPUOCKGR-UHFFFAOYSA-N
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
11704
tetraethylstannane
CC[Sn](CC)(CC)CC
This item is not returnable. View return policy
597-64-8
C8H20Sn
234.96
MFCD00009020
RWWNQEOPUOCKGR-UHFFFAOYSA-N
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
11704
tetraethylstannane
CC[Sn](CC)(CC)CC

Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -112°C
Density 1.187g/mL
Boiling Point 181°C
Flash Point 53°C (128°F)
Odor Characteristic
Refractive Index 1.4730
Quantity 1 g
UN Number UN3384
Solubility Information Insoluble in water; Soluble in alcohol and ether
Formula Weight 234.94
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetraethyltin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'