missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethyltin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
Description
Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -112°C |
Density | 1.187 g/mL |
Boiling Point | 181°C |
Flash Point | 53°C (128°F) |
Odor | Characteristic |
Refractive Index | 1.473 |
Quantity | 50 g |
UN Number | UN3384 |
Solubility Information | Insoluble in water; Soluble in alcohol and ether |
Formula Weight | 234.94 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'