missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylpropylene-1,3-diphosphonate, 97%, Thermo Scientific™

Catalog Number 10205631
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS 22401-25-8
Color Colorless
Boiling Point 206.0°C to 227.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H26O6P2
Refractive Index 1.4450 to 1.4470
MDL Number MFCD00048543
Quantity 5 g
Synonym phosphonic acid, trimethylenedi-, tetraethyl ester, phosphonic acid, 1,3-propanediylbis-, tetraethyl ester, tetraethyl propylenediphosphonate, tetraethyltrimethylenediphosphonate, 1,3-bis diethoxyphosphinyl propane, 1,3-bis diethoxyphosphoryl propane, tetraethyl propane-1,3-diphosphonate, tetraethyl 1,3-propylene bisphosphonate, propanediyl-bis-phosphonic acid tetraethyl ester, propane-1,3-diphosphonic acid, tetraethyl ester
InChI Key LJQJJGQRJQSYQN-UHFFFAOYSA-N
SMILES CCOP(=O)(CCCP(=O)(OCC)OCC)OCC
IUPAC Name 1,3-bis(diethoxyphosphoryl)propane
Molecular Weight (g/mol) 316.27
PubChem CID 519945
Formula Weight 316.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tetraethylpropylene-1, 3-diphosphonate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'