missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol, 99.5%, Thermo Scientific Chemicals

Catalog Number p-4510151
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
112-60-7
C8H18O5
194.23
UWHCKJMYHZGTIT-UHFFFAOYSA-N
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
8200
CHEBI:44920
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
C(COCCOCCOCCO)O
112-60-7
C8H18O5
194.23
UWHCKJMYHZGTIT-UHFFFAOYSA-N
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
8200
CHEBI:44920
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
C(COCCOCCOCCO)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -4.1°C
Color Yellow
Density 1.1200g/mL
Boiling Point 324.0°C to 330.0°C
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass Bottle
Linear Formula O(CH2CH2OCH2CH2OH)2
Refractive Index 1.4580 to 1.4600
Quantity 1 L
Beilstein 01,468
Specific Gravity 1.12
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene and toluene, practically insoluble in diethylether
Viscosity 44.9 mPa.s (25°C)
Formula Weight 194.23
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'