missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water, Thermo Scientific Chemicals
10.02 GBP valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Use promo code "21636" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 10 g |
UN Number | UN1479 |
Beilstein | 3638093 |
Solubility Information | Soluble in water,acetone,acetonitrile and dimethyl sulfoxide. |
Formula Weight | 229.71 |
Percent Purity | 98% |
Concentration or Composition (by Analyte or Components) | Contains ≈ 10% water |
Physical Form | Wet Crystals |
Show More |
Safety and Handling
Product Identifier
- Tetraethlammonium perchlorate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P362-Take off contaminated clothing.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'