Learn More
Tetraethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 420295000
Code : 29
Additional Details : CAS Number : 77-98-5 Weight : 0.50000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetraethylammonium hydroxide | |
25% in Water | |
102.0°C | |
(C2H5)4NOH | |
1.0200g/mL | |
Solubility in water: soluble. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Plastic bottle | |
1.02 | |
Solution |
Colorless to Yellow | |
77-98-5,7732-18-5 | |
C8H21NO | |
MFCD00009024 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
24 to 26% | |
1.402 | |
500 g |
RUO – Research Use Only