missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydrogensulfate, 99+%, HPLC grade, Thermo Scientific Chemicals

Catalog Number p-3830594
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16873-13-5
C8H21NO4S
227.32
MFCD00036150
CREVBWLEPKAZBH-UHFFFAOYSA-M
tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate
167582
hydrogen sulfate;tetraethylazanium
OS([O-])(=O)=O.CC[N+](CC)(CC)CC
This item is not returnable. View return policy
16873-13-5
C8H21NO4S
227.32
MFCD00036150
CREVBWLEPKAZBH-UHFFFAOYSA-M
tetraethylammonium hydrogensulfate, tetraethylammonium hydrogen sulphate, tetraethylammonium hydrogen sulfate, n,n,n-triethylethanaminium hydrogen sulfate 1:1:1, hydrogen sulfate; tetraethylazanium, tertraethylammonium hydrogen sulfate, tetraethyl ammonium hydrogen sulphate
167582
hydrogen sulfate;tetraethylazanium
OS([O-])(=O)=O.CC[N+](CC)(CC)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Absorbance 0.02 max. (0.4 M in HPLC water) at 250nm, 0.04 max. (0.4 M in HPLC water) at 230nm, 0.04 max. (0.4 M in HPLC water) at 240nm, 0.05 max. (0.4 M in HPLC water) at 220nm, 0.1 max. (0.4 M in HPLC water) at 210nm
Melting Point 243°C to 245°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Formula Weight 227.32
Percent Purity 99+%
Grade HPLC
Physical Form Powder
Chemical Name or Material Tetraethylammonium hydrogensulfate
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium hydrogensulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'