missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium fluoride hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-3830292
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
98330-04-2
C8H20FN
149.25
MFCD00149990
QSUJAUYJBJRLKV-UHFFFAOYSA-M
tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate
16211652
[F-].CC[N+](CC)(CC)CC
This item is not returnable. View return policy
98330-04-2
C8H20FN
149.25
MFCD00149990
QSUJAUYJBJRLKV-UHFFFAOYSA-M
tetraethylammonium fluoride hydrate, tetraethylammonium hydrate fluoride, tetraethylammoniumfluoridehydrate, acmc-209nvn, et4nf.h2o, tetraethylazanium fluoride hydrate, tetraethyl ammonium fluoride hydrate, ethanaminium, n,n,n-triethyl-, fluoride, monohydrate
16211652
[F-].CC[N+](CC)(CC)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5)4NF·xH2O
Quantity 5 g
IUPAC Name tetraethylazanium;fluoride;hydrate
Formula Weight 149.14
Percent Purity 98%
Physical Form Adhering Crystals, Crystals or Crystalline Solid
Chemical Name or Material Tetraethylammonium fluoride hydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetraethylammonium fluoride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'