missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog Number 10792771
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
71-91-0
C8H20BrN
210.16
MFCD00011825
HWCKGOZZJDHMNC-UHFFFAOYSA-M
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
6285
tetraethylazanium;bromide
CC[N+](CC)(CC)CC.[Br-]
71-91-0
C8H20BrN
210.16
MFCD00011825
HWCKGOZZJDHMNC-UHFFFAOYSA-M
tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
6285
tetraethylazanium;bromide
CC[N+](CC)(CC)CC.[Br-]

Phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 285.0°C
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (C2H5)4NBr
Quantity 250 g
Beilstein 04,104
Fieser 06,568; 13,46
Merck Index 15,9344
Solubility Information Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol,chloroform and acetone,slightly soluble in benzene
Formula Weight 210.16
Percent Purity 98%
Chemical Name or Material Tetraethylammonium bromide, 98%
Show More Show Less
Product Identifier
  • Tetraethylammonium bromide
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'