missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethoxysilane, 98%, Thermo Scientific Chemicals
15.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
Solubility
Soluble in ethanol and 2-propanol.
Notes
Degrades in water. Keep container tightly closed in a dry and well-ventilated place.
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.
Solubility
Soluble in ethanol and 2-propanol.
Notes
Degrades in water. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -85°C |
Density | 0.934 |
Boiling Point | 168°C |
Flash Point | 46°C (116°F) |
Assay Percent Range | 98% |
Odor | Pleasant |
Linear Formula | (C2H5O)4Si |
Refractive Index | 1.383 |
Quantity | 100 mL |
UN Number | UN1292 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'