missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Ultrapure, Spectrophotometric Grade, 99+%, Thermo Scientific Chemicals

Catalog Number p-4408478
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
ClC(Cl)=C(Cl)Cl
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
ClC(Cl)=C(Cl)Cl

Tetrachloroethylene is used as solvent. It is involved in dry cleaning as well as in the degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 22°C
Density 1.623 g/mL
Boiling Point 120°C to 121°C
Assay Percent Range >99%
Refractive Index 1.5055
Quantity 1 L
Beilstein 1361721
Solubility Information Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water.
Formula Weight 165.83
Grade Spectrophotometric
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene, Ultrapure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'