missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3737449
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
ClC(Cl)=C(Cl)Cl
127-18-4
C2Cl4
165.82
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
ClC(Cl)=C(Cl)Cl

Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 2.5L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Alkalinity 0.003% max. (NaOH)
Melting Point -22.0°C
Density 1.6250g/mL
Boiling Point 120.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula Cl2C=CCl2
Refractive Index 1.505
Quantity 500 mL
Beilstein 01, 187
Residue after Evaporation 0.005% max.
Merck Index 15, 9335
Specific Gravity 1.625
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene
Viscosity 0.89 mPa.s (20°C)
Formula Weight 165.83
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrachloroethylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'