missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals

Catalog Number 11314517
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
31011-57-1
C8H16Cl4O2Ti
333.881
MFCD00077884
LXWBMENBONGPSB-UHFFFAOYSA-J
tetrachlorobis tetrahydrofuran titanium, tetrachlorobis tetrahydrofuran titanium iv, bis tetrahydrofuran ; titanium tetrachloride, titanium iv chloride tetrahydrofuran complex
10958563
oxolane;tetrachlorotitanium
C1CCOC1.C1CCOC1.Cl[Ti](Cl)(Cl)Cl
This item is not returnable. View return policy
31011-57-1
C8H16Cl4O2Ti
333.881
MFCD00077884
LXWBMENBONGPSB-UHFFFAOYSA-J
tetrachlorobis tetrahydrofuran titanium, tetrachlorobis tetrahydrofuran titanium iv, bis tetrahydrofuran ; titanium tetrachloride, titanium iv chloride tetrahydrofuran complex
10958563
oxolane;tetrachlorotitanium
C1CCOC1.C1CCOC1.Cl[Ti](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 128°C
Color Yellow
Flash Point −17° (1°F)
Quantity 50 g
UN Number UN2925
Sensitivity Moisture Sensitive
Formula Weight 333.91
Percent Purity 98%
Physical Form Crystalline or Powder
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'