missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetrachlorobis(tetrahydrofuran)niobium(IV)

Brand:  Alfa Aesar™ 039460.04

Code : NEW

Additional Details : CAS Number : 61247-57-2 Weight : 0.00200kg Transport : UN number : 2925 Chem class : 1 Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11398258

  • £38.37 / 2g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Tetrachlorobis(tetrahydrofuran)niobium(IV)
378.94
UN2925
61247-57-2
MFCD00013405
Niobium(IV) chloride - tetrahydrofuran complex

Specifications

Tetrachlorobis(tetrahydrofuran)niobium(IV)
Powder
MFCD00013405
Niobium(IV) chloride - tetrahydrofuran complex
93°C (dec.)
2g
61247-57-2
378.94
UN2925
Yellow-orange to brown
49° (120°F)
Moisture Sensitive
Videos
SDS