missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloro-o-benzoquinone, 97%, Thermo Scientific Chemicals

Catalog Number 10183571
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2435-53-2
C6Cl4O2
245.86
MFCD00001646
VRGCYEIGVVTZCC-UHFFFAOYSA-N
o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
73252
3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione
ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl
2435-53-2
C6Cl4O2
245.86
MFCD00001646
VRGCYEIGVVTZCC-UHFFFAOYSA-N
o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
73252
3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione
ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 125.0°C to 129.0°C
Color Bordeaux
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6Cl4(=O)2
Quantity 5 g
Beilstein 07, 602
Fieser 01,128; 04,76; 05,104; 07,355; 09,102; 10,85
Solubility Information Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C)
Formula Weight 245.88
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrachloro-o-benzoquinone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrachloro-o-benzoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'