missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloro-o-benzoquinone, 97%, Thermo Scientific Chemicals
107.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 125.0°C to 129.0°C |
Color | Bordeaux |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C6Cl4(=O)2 |
Quantity | 25 g |
Beilstein | 07, 602 |
Fieser | 01,128; 04,76; 05,104; 07,355; 09,102; 10,85 |
Solubility Information | Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C) |
Show More |
Safety and Handling
Product Identifier
- Tetrachloro-o-benzoquinone
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'