missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium phosphate monobasic, 99%, HPLC grade, Thermo Scientific Chemicals

Catalog Number p-195853
59.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5574-97-0
C16H38NO4P
339.46
MFCD00064526
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
2735142
dihydrogen phosphate;tetrabutylazanium
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5574-97-0
C16H38NO4P
339.46
MFCD00064526
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
2735142
dihydrogen phosphate;tetrabutylazanium
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Absorbance (0.1% aq. soln.) (1 cm quartz cell), (0.1% aq. soln.) (1 cm quartz cell) at 0nm, 0.05 max. at 214nm, 0.1 max. at 210nm, 0.2 max. at 200nm
Melting Point 148.0°C to 151.0°C
Color Gray to Black
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3(CH2)3)4NH2PO4
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 339.45
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tetrabutylammonium phosphate monobasic, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'