missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide Solution, 0.1M, Honeywell Fluka™

Catalog Number p-7072282
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
2052-49-5
C16H37NO
259.48
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
[OH-].CCCC[N+](CCCC)(CCCC)CCCC

Volumetric, 0.1 M (C4H9)4NOH (isopropanol/methanol (10:1 V/V)), for titration in non-aqueous medium

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Flash Point 15°C (59°F)
Linear Formula (CH3CH2CH2CH2)4N(OH)
Quantity 250 mL
UN Number UN1992
Beilstein 3571228
IUPAC Name tetrabutylazanium;hydroxide
Formula Weight 259.47g/mol
Chemical Name or Material Tetrabutylammonium hydroxide solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'