Learn More
Tetrabutylammonium Hydrogen Sulfate, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical T/0479/48
Code : 21
Additional Details : CAS Number : 32503-27-8 Weight : 0.10000kg
Quantity | 100g |
---|
Chemical Identifiers
32503-27-8 | |
339.535 | |
SHFJWMWCIHQNCP-UHFFFAOYSA-M | |
94433 | |
CCCC[N+](CCCC)(CCCC)CCCC.OS(=O)(=O)[O-] |
C16H37NO4S | |
11637 | |
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs | |
hydrogen sulfate;tetrabutylazanium |
Specifications
Plastic powder jar | |
168°C | |
2.1 | |
C16H37NO4S | |
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs | |
CCCC[N+](CCCC)(CCCC)CCCC.OS(=O)(=O)[O-] | |
339.535 | |
339.53g/mol |
White | |
100g | |
32503-27-8 | |
11637 | |
SHFJWMWCIHQNCP-UHFFFAOYSA-M | |
hydrogen sulfate;tetrabutylazanium | |
94433 | |
Solid |
Safety and Handling
- Tetrabutylammonium hydrogen sulfate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Acute toxicity Category 4
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.