missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium fluoride, 75% solution in water, Thermo Scientific Chemicals

Catalog Number 15254904
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
429-41-4
C16H36FN
261.47
MFCD00011747
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
[F-].CCCC[N+](CCCC)(CCCC)CCCC
This item is not returnable. View return policy
429-41-4
C16H36FN
261.47
MFCD00011747
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
[F-].CCCC[N+](CCCC)(CCCC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9530g/mL
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NF
Quantity 500 g
Merck Index 15,9332
Specific Gravity 0.953
IUPAC Name tetrabutylazanium;fluoride
Formula Weight 261.46
Chemical Name or Material Tetrabutylammonium fluoride
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium fluoride, 75% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'